AC5372911315 Accela ChemBio Inc.

Ethyl 3-Hydroxy-3-phenylpropanoate

  • MDL number MFCD00086891
  • Cas Number : 5764-85-2
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015621&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY015621.pdf
hstatements H315-H318-H335
Availability details for
Quantity
  • Availability
  • Add to Cart