AC5372911318 Accela ChemBio Inc.

Ethyl (R)-1-Boc-3-pyrrolidinecarboxylate

  • MDL number MFCD19442798
  • Cas Number : 1314419-65-2
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015625&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280-P305+P351+P338-P304+P340-P405-P501A
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY015625.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart