AC5372911320 Accela ChemBio Inc.

Ethyl Imidazo[1,2-a]pyridine-7-carboxylate

  • MDL number MFCD08436046
  • Cas Number : 372147-49-4
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015645&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015645.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart