AC5372911345 Accela ChemBio Inc.

Methyl 2-Hexenoate

  • MDL number MFCD00048798
  • Cas Number : 2396-77-2
  • Molecular Weight : 128.16898

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015809&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015809.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart