AC5372911347 Accela ChemBio Inc.

9,10-Bis(4-methoxyphenyl)-2-chloroanthracene

  • MDL number MFCD00009935
  • Cas Number : 110904-87-5
  • Molecular Weight : 424.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY015812&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY015812.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart