AC5372911376 Accela ChemBio Inc.

Ethyl 2-Chloro-4,6-dihydroxynicotinate

  • MDL number MFCD12546397
  • Cas Number : 70180-38-0
  • Molecular Weight : 217.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016051&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016051.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart