AC5372911378 Accela ChemBio Inc.

4-Fluorophenyl Acetate

  • MDL number MFCD00041361
  • Cas Number : 405-51-6
  • Molecular Weight : 154.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016072&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P280-P403+P235-P501
hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY016072.pdf
hstatements H227
Availability details for
Quantity
  • Availability
  • Add to Cart