AC5372911380 Accela ChemBio Inc.

Ethyl Thiomorpholine-2-carboxylate

  • MDL number MFCD20134212
  • Cas Number : 152009-44-4
  • Molecular Weight : 175.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016104&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016104.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart