AC5372911381 Accela ChemBio Inc.

Ethyl N-Boc-2-thiomorpholinecarboxylate

  • MDL number MFCD20134213
  • Cas Number : 1346597-50-9
  • Molecular Weight : 275.36444

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016105&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016105.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart