AC5372911389 Accela ChemBio Inc.

2-Hydroxytetrahydrofuran

  • MDL number MFCD07369457
  • Cas Number : 5371-52-8
  • Molecular Weight : 88.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form Colorless to yellow liquid
Shipping Temperature +4 ?
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016132&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY016132.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart