AC5372911412 Accela ChemBio Inc.

N-(2,3-Dimethyl-5-oxo-1-phenyl-2,5-dihydro-1H-pyrazol-4-yl)-2,2,2-trifluoroacetamide

  • MDL number MFCD00574858
  • Cas Number : 129476-61-5
  • Molecular Weight : 299.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016214&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016214.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart