AC5372911417 Accela ChemBio Inc.

1-(Boc-amino)-3-bromo-1-(4-methoxyphenyl)propane

  • MDL number MFCD06410617
  • Cas Number : 924818-01-9
  • Molecular Weight : 344.24408

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016247&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016247.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart