AC5372911420 Accela ChemBio Inc.

3-Amino-3-(4-methylphenyl)propionic Acid

  • MDL number MFCD01863249
  • Cas Number : 68208-18-4
  • Molecular Weight : 179.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016253&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016253.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart