AC5372911427 Accela ChemBio Inc.

1-Benzyl-3-(chloromethyl)-1H-indazole

  • MDL number MFCD20134218
  • Cas Number : 131427-22-0
  • Molecular Weight : 256.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016298&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016298.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart