AC5372911428 Accela ChemBio Inc.

Benzyl 1-Benzyl-1H-indazole-3-carboxylate

  • MDL number MFCD20134219
  • Cas Number : 174180-54-2
  • Molecular Weight : 342.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016300&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016300.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart