AC5372911444 Accela ChemBio Inc.

4-Nitroimidazole-5-carbonitrile

  • MDL number MFCD19218753
  • Cas Number : 208510-03-6
  • Molecular Weight : 138.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016355&attname=GoodsCode

Safety Information

signalword Warning
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY016355.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart