AC5372911446 Accela ChemBio Inc.

1-Cbz-3-(bromomethyl)piperidine

  • MDL number MFCD06410596
  • Cas Number : 242459-81-0
  • Molecular Weight : 312.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016375&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016375.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart