AC5372911476 Accela ChemBio Inc.

Ethyl 3-Morpholinecarboxylate

  • MDL number MFCD12406992
  • Cas Number : 84005-98-1
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016483&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016483.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart