AC5372911486 Accela ChemBio Inc.

Ethyl 4-(3-Aminophenyl)-5-(methoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-3-carboxylate

  • MDL number MFCD20491402
  • Cas Number : 138135-48-5
  • Molecular Weight : 330.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016507&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016507.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart