AC5372911487 Accela ChemBio Inc.

4-Amino-3-methyl-N-(4-methyl-2-pyridyl)benzamide

  • MDL number MFCD11131807
  • Cas Number : 1019398-78-7
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016508&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016508.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart