AC5372911488 Accela ChemBio Inc.

3-Amino-4-chloro-N-(3-pyridylmethyl)benzamide

  • MDL number MFCD10021697
  • Cas Number : 1018502-06-1
  • Molecular Weight : 261.70688

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016509&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016509.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart