AC5372911490 Accela ChemBio Inc.

5-Amino-2-chloro-N-(2-furylmethyl)benzamide

  • MDL number MFCD09044284
  • Cas Number : 926216-59-3
  • Molecular Weight : 250.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016514&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016514.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart