AC5372911497 Accela ChemBio Inc.

2-Bromo-5-methylhexane

  • MDL number MFCD07780428
  • Cas Number : 6570-93-0
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016753&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY016753.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart