AC5372911522 Accela ChemBio Inc.

(1S,6R)-3-Boc-3,7-diazabicyclo[4.2.0]octane

  • MDL number MFCD21332889
  • Cas Number : 1417789-49-1
  • Molecular Weight : 212.2887

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. -20?
Physical Form solid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016881&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016881.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart