AC5372911537 Accela ChemBio Inc.

2-[2-(Cbz-amino)phenyl]-1,3-dioxolane

  • MDL number MFCD20491415
  • Cas Number : 1346597-58-7
  • Molecular Weight : 299.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016920&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016920.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart