AC5372911538 Accela ChemBio Inc.

Ethyl 3-tert-Butyl-1-methylpyrazole-5-carboxylate

  • MDL number MFCD00085059
  • Cas Number : 133261-10-6
  • Molecular Weight : 210.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016923&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016923.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart