AC5372911550 Accela ChemBio Inc.

(S)-1-[(R)-1-Phenylethyl]-3-(hydroxymethyl)pyrrolidine

  • MDL number MFCD12828252
  • Cas Number : 109960-55-6
  • Molecular Weight : 205.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016947&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016947.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart