AC5372911554 Accela ChemBio Inc.

Ethyl 2-(N-Methoxy-N-methylamino)-2-oxoacetate

  • MDL number MFCD11975566
  • Cas Number : 139507-52-1
  • Molecular Weight : 161.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016958&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016958.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart