AC5372911558 Accela ChemBio Inc.

4-(Cbz-amino)-1-(methylsulfonyl)piperidine

  • MDL number MFCD20491416
  • Cas Number : 402927-96-2
  • Molecular Weight : 312.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY016981&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY016981.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart