AC5372911573 Accela ChemBio Inc.

4-(4-Bromophenyl)tetrahydropyran

  • MDL number MFCD12913643
  • Cas Number : 811828-76-9
  • Molecular Weight : 241.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017017&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017017.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart