AC5372911576 Accela ChemBio Inc.

2,6-Dichloro-3-nitrobenzyl Bromide

  • MDL number MFCD20491417
  • Cas Number : 83141-02-0
  • Molecular Weight : 284.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017028&attname=GoodsCode

Safety Information

signalword Danger
hcodes GHS05, GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY017028.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart