AC5372911600 Accela ChemBio Inc.

2-Butyl-4-chloroquinoline

  • MDL number MFCD20491419
  • Cas Number : 144624-27-1
  • Molecular Weight : 219.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017127&attname=GoodsCode

Safety Information

hcodes No data known
MSDS http://pdf.accelachem.com/cn/msds/SY017127.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart