AC5372911609 Accela ChemBio Inc.

4,5-Dimethoxy-2-methylbenzoic Acid

  • MDL number MFCD00029889
  • Cas Number : 20736-28-1
  • Molecular Weight : 196.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017157&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017157.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart