AC5372911612 Accela ChemBio Inc.

Methyl 4-Fluoro-3-(methylsulfonyl)benzoate

  • MDL number MFCD20491420
  • Cas Number : 160819-39-6
  • Molecular Weight : 232.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017163&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017163.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart