AC5372911622 Accela ChemBio Inc.

5-Nitrobarbituric Acid

  • MDL number MFCD00006667
  • Cas Number : 480-68-2
  • Molecular Weight : 173.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017190&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY017190.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart