AC5372911657 Accela ChemBio Inc.

2-(2,4-Dichlorophenoxy)-1-(1-piperidyl)-1-propanone

  • MDL number MFCD01339369
  • Cas Number : 101991-76-8
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017416&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017416.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart