AC5372911660 Accela ChemBio Inc.

N-(2-Hydroxypropyl)-4-methylbenzenesulfonamide

  • MDL number MFCD02225949
  • Cas Number : 59724-53-7
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017419&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017419.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart