AC5372911663 Accela ChemBio Inc.

Boc-D-aspartinol 4-Benzyl Ester

  • MDL number MFCD02094135
  • Cas Number : 182748-72-7
  • Molecular Weight : 309.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017424&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017424.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart