AC5372911717 Accela ChemBio Inc.

8-Methyl-2-oxo-1,2,3,4-tetrahydroquinoline-6-boronic Acid Pinacol Ester

  • MDL number MFCD22423111
  • Molecular Weight : 287.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY020007&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY020007.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart