AC5372911719 Accela ChemBio Inc.

2,4-Diethylpyrimidine

  • MDL number MFCD22423156
  • Cas Number : 857990-28-4
  • Molecular Weight : 136.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,4-diethylpyrimidine
Inchi Code 1S/C8H12N2/c1-3-7-5-6-9-8(4-2)10-7/h5-6H,3-4H2,1-2H3
Inchi Key LOACNDPOIZYNHB-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021214&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021214.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart