AC5372911720 Accela ChemBio Inc.

1-(Boc-aminomethyl)-3-(5-tetrazolyl)benzene

  • MDL number MFCD22423157
  • Cas Number : 1398534-55-8
  • Molecular Weight : 275.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021217&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021217.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart