AC5372911723 Accela ChemBio Inc.

Methyl 2-(Cbz-amino)-2-(tetrahydrothiopyran-4-ylidene)acetate

  • MDL number MFCD22574985
  • Cas Number : 894790-18-2
  • Molecular Weight : 321.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021519&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY021519.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart