AC5372911724 Accela ChemBio Inc.

4-(1-Benzyl-4-piperidylidene)-2-buten-1-ol

  • MDL number MFCD22574987
  • Cas Number : 1408229-29-7
  • Molecular Weight : 243.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2E)-4-(1-benzyl-4-piperidinylidene)-2-buten-1-ol
Inchi Code 1S/C16H21NO/c18-13-5-4-6-15-9-11-17(12-10-15)14-16-7-2-1-3-8-16/h1-8,18H,9-14H2/b5-4+
Inchi Key WHNOLOCULOSWCX-SNAWJCMRSA-N

Description

Purity 95%
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY021556&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY021556.pdf
hcodes NO GHS SYMBOL
Availability details for
Quantity
  • Availability
  • Add to Cart