AC5372911731 Accela ChemBio Inc.

tert-Butyl (R)-6-Cyano-5-hydroxy-3-oxohexanoate

  • MDL number MFCD16251319
  • Cas Number : 125988-01-4
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017595&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017595.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart