AC5372911732 Accela ChemBio Inc.

tert-Butyl 2-[(4R,6S)-6-(Acetoxymethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate

  • MDL number MFCD08458213
  • Cas Number : 154026-95-6
  • Molecular Weight : 302.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017597&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017597.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart