AC5372911739 Accela ChemBio Inc.

Ethyl 2-Fluoro-5-methylbenzoate

  • MDL number MFCD06204763
  • Cas Number : 496841-90-8
  • Molecular Weight : 182.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017650&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017650.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart