AC5372911767 Accela ChemBio Inc.

4-Bromo-N-(5-chloro-2-pyridyl)benzamide

  • MDL number MFCD00436765
  • Cas Number : 300670-29-5
  • Molecular Weight : 311.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY017943&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY017943.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart