AC5372911806 Accela ChemBio Inc.

1-Boc-4-[2-(aminomethyl)phenyl]piperazine

  • MDL number MFCD04115066
  • Cas Number : 174855-53-9
  • Molecular Weight : 291.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20?
Physical Form liquid
Shipping Temperature +4 ?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018126&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P330+P331-P280-P305+P351+P338-P310-P301+P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018126.pdf
hstatements H318-H314-H302
Availability details for
Quantity
  • Availability
  • Add to Cart