AC5372911827 Accela ChemBio Inc.

2-(2,3-Dihydrobenzofuran-5-yl)-N-(5-methylisoxazol-3-yl)acetamide

  • MDL number MFCD21364422
  • Cas Number : 1363166-39-5
  • Molecular Weight : 258.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018193&attname=GoodsCode

Safety Information

hcodes NO GHS SYMBOL
MSDS http://pdf.accelachem.com/cn/msds/SY018193.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart