AC5372911836 Accela ChemBio Inc.

(R)-3-Amino-1-benzylpiperidine

  • MDL number MFCD03093374
  • Cas Number : 168466-84-0
  • Molecular Weight : 190.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY018216&attname=GoodsCode

Safety Information

signalword Danger
pcodes P301+P310
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY018216.pdf
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart